Assay Details
| Gene Symbol: NCK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257480.1 | NP_001244409.1 | 1054 | 110 | ||
| XM_015132050.2 | XP_014987536.1 | 1095 | 110 | |||
| XM_015132051.2 | XP_014987537.1 | 1090 | 110 | |||
| XM_015132052.2 | XP_014987538.1 | 1046 | 110 | |||
| XM_015132053.2 | XP_014987539.1 | 1305 | 110 | |||
| XM_028843422.1 | XP_028699255.1 | 1174 | 110 | |||
| XM_028843423.1 | XP_028699256.1 | 1133 | 110 | |||
| XM_028843424.1 | XP_028699257.1 | 975 | 110 | |||
| GenBank mRNA | JU320326.1 | - | 1054 | 110 | ||
| JU475697.1 | - | 1008 | 110 | |||
| JV047145.1 | - | 954 | 110 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

NCK1
Genomic Map