Assay Details
| Gene Symbol: GLRX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266398.1 | NP_001253327.1 | 401 | 106 | ||
| XM_015121032.2 | XP_014976518.1 | 1094 | 106 | |||
| XM_028844650.1 | XP_028700483.1 | 489 | 106 | |||
| GenBank mRNA | JU334762.1 | - | 401 | 106 | ||
| JU487611.1 | - | 255 | 106 | |||
| JV508615.1 | - | 288 | 106 | |||
More Information
Set Membership: |
|

GLRX2
Genomic Map