Assay Details
Gene Symbol: TNFAIP3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261344.1 | NP_001248273.1 | 1051 | 117 | ||
XM_015137275.2 | XP_014992761.1 | 2706 | 117 | |||
GenBank mRNA | JU470469.1 | - | 1051 | 117 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
