Assay Details
| Gene Symbol: FAM76A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001111916.4 | XP_001111916.1 | 88 | |||
| XM_001111995.4 | XP_001111995.1 | 88 | ||||
| GenBank mRNA | JU320723.1 | - | 88 | |||
| JU320724.1 | - | 88 | ||||
| JV044953.1 | - | 215 | 88 | |||
More Information
Set Membership: |
|

FAM76A
Genomic Map