Assay Details
| Gene Symbol: RAD51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143966.2 | XP_014999452.2 | 1236 | 98 | ||
| GenBank mRNA | JU325731.1 | - | 40 | 98 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RAD51B| Gene Symbol: RAD51B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015143966.2 | XP_014999452.2 | 1236 | 98 | ||
| GenBank mRNA | JU325731.1 | - | 40 | 98 | ||
Set Membership: |
|