Assay Details
Gene Symbol: LOC713572 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143401.2 | XP_014998887.1 | 953 | 90 | ||
XM_015143402.2 | XP_014998888.1 | 1006 | 90 | |||
XM_028851425.1 | XP_028707258.1 | 595 | 90 | |||
GenBank mRNA | JU319755.1 | - | 625 | 90 | ||
JV043570.1 | - | 619 | 90 | |||
JV636544.1 | - | 633 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
