Assay Details
| Gene Symbol: MYD88 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001130681.1 | NP_001124153.1 | 739 | 94 | ||
| GenBank mRNA | AB446476.1 | - | 739 | 94 | ||
| EU204914.1 | - | 751 | 94 | |||
| JU473229.1 | - | 801 | 94 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MYD88
Genomic Map