Assay Details
| Gene Symbol: LOC100425407 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844189.1 | XP_028700022.1 | 994 | 127 | ||
| GenBank mRNA | JU323020.1 | - | 980 | 127 | ||
| JU472825.1 | - | 966 | 127 | |||
| JV044383.1 | - | 926 | 127 | |||
| JV487714.1 | - | 162 | 127 | |||
| Gene Symbol: LOC698851 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028844191.1 | XP_028700024.1 | 1747 | 127 | ||
More Information
Set Membership: |
|

LOC100425407
Genomic Map