Assay Details
| Gene Symbol: DYNLRB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257686.1 | NP_001244615.1 | 355 | 147 | ||
| XM_028840572.1 | XP_028696405.1 | 329 | 147 | |||
| GenBank mRNA | JU320704.1 | - | 355 | 147 | ||
| JU472814.1 | - | 303 | 147 | |||
| JV045418.1 | - | 254 | 147 | |||
More Information
Set Membership: |
|

DYNLRB2
Genomic Map