Assay Details
| Gene Symbol: PLPPR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015145652.2 | XP_015001138.1 | 690 | 75 | ||
| GenBank mRNA | JU327604.1 | - | 368 | 75 | ||
| JV566102.1 | - | 566 | 75 | |||
More Information
Set Membership: |
|

PLPPR4
Genomic Map