Assay Details
| Gene Symbol: ISLR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001097240.3 | XP_001097240.2 | 344 | 66 | ||
| GenBank mRNA | JV046982.1 | - | 265 | 66 | ||
| JV637061.1 | - | 278 | 66 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001097240.3 transcripts.
Set Membership: |
|

ISLR
Genomic Map