Assay Details
| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115222.2 | XP_014970708.2 | 1022 | 65 | ||
| XM_015115223.2 | XP_014970709.1 | 1022 | 65 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MAP6| Gene Symbol: MAP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015115222.2 | XP_014970708.2 | 1022 | 65 | ||
| XM_015115223.2 | XP_014970709.1 | 1022 | 65 | |||
Set Membership: |
|