Assay Details
| Gene Symbol: KBTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266403.1 | NP_001253332.1 | 2224 | 94 | ||
| GenBank mRNA | JU336978.1 | - | 2224 | 94 | ||
| JV597561.1 | - | 2205 | 94 | |||
More Information
Set Membership: |
|

KBTBD6
Genomic Map