Assay Details
| Gene Symbol: CYP4V2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001193838.2 | NP_001180767.2 | 622 | 104 | ||
| XM_015139562.2 | XP_014995048.2 | 217 | 104 | |||
| XM_028847981.1 | XP_028703814.1 | 129 | 104 | |||
| GenBank mRNA | JU322982.1 | - | 362 | 104 | ||
More Information
Set Membership: |
|

CYP4V2
Genomic Map