Assay Details
| Gene Symbol: WT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015114575.2 | XP_014970061.1 | 1527 | 78 | ||
| XM_015114576.2 | XP_014970062.1 | 1527 | 78 | |||
| XM_015114577.2 | XP_014970063.1 | 1386 | 78 | |||
| XM_028833532.1 | XP_028689365.1 | 980 | 78 | |||
| XM_028833533.1 | XP_028689366.1 | 980 | 78 | |||
| XM_028833534.1 | XP_028689367.1 | 934 | 78 | |||
| XM_028833535.1 | XP_028689368.1 | 721 | 78 | |||
| GenBank mRNA | JV048171.1 | - | 1372 | 78 | ||
| JV048172.1 | - | 1423 | 78 | |||
| JV048173.1 | - | 1423 | 78 | |||
| JV660606.1 | - | 1468 | 78 | |||
| JV660607.1 | - | 1519 | 78 | |||
More Information
Set Membership: |
|

WT1
Genomic Map