Assay Details
| Gene Symbol: SLX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001194449.2 | NP_001181378.1 | 1006 | 119 | ||
| GenBank mRNA | JU470693.1 | - | 1021 | 119 | ||
| JV635534.1 | - | 921 | 119 | |||
More Information
Set Membership: |
|

SLX1A
Genomic Map