Assay Details
| Gene Symbol: GZMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_001114420.4 | XP_001114420.3 | 176 | 66 | ||
| GenBank mRNA | JU475347.1 | - | 113 | 66 | ||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

GZMB
Genomic Map