Assay Details
| Gene Symbol: CD40LG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032839.1 | NP_001028011.1 | 382 | 135 | ||
| GenBank mRNA | AF344859.1 | - | 382 | 135 | ||
| JU471432.1 | - | 401 | 135 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD40LG
Genomic Map