Assay Details
| Gene Symbol: ICAM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001047135.1 | NP_001040600.1 | 1427 | 121 | ||
| GenBank mRNA | AF340040.1 | - | 1427 | 121 | ||
| JU475417.1 | - | 1469 | 121 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

ICAM1
Genomic Map