Assay Details
| Gene Symbol: CCL19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032959.1 | NP_001028131.1 | 49 | 60 | ||
| GenBank mRNA | AF449273.1 | - | 49 | 60 | ||
| JU491287.1 | - | 76 | 60 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CCL19
Genomic Map