Assay Details
| Gene Symbol: CD86 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001042644.2 | NP_001036109.1 | 401 | 120 | ||
| GenBank mRNA | AF344857.1 | - | 401 | 120 | ||
| JU477900.1 | - | 513 | 120 | |||
| JV704569.1 | - | 499 | 120 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

CD86
Genomic Map