Assay Details
| Gene Symbol: MAMU-DRA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001134298.1 | NP_001127770.1 | 80 | 151 | ||
| XM_015135912.2 | XP_014991398.2 | 184 | 151 | |||
| GenBank mRNA | AJ586874.1 | - | 80 | 151 | ||
| AJ586875.1 | - | 80 | 151 | |||
| AJ586876.1 | - | 80 | 151 | |||
| AJ586877.1 | - | 80 | 151 | |||
| AJ586878.1 | - | 80 | 151 | |||
| AJ586880.1 | - | 80 | 151 | |||
| AJ586881.1 | - | 80 | 151 | |||
| AJ586882.1 | - | 80 | 151 | |||
| AJ586883.1 | - | 80 | 151 | |||
| AJ586884.1 | - | 80 | 151 | |||
| EU934772.1 | - | 151 | ||||
| GQ471896.1 | - | 80 | 151 | |||
| HF586896.1 | - | 80 | 151 | |||
| HF586897.1 | - | 80 | 151 | |||
| HF586898.1 | - | 80 | 151 | |||
| HF586899.1 | - | 80 | 151 | |||
| HF586900.1 | - | 80 | 151 | |||
| HF586901.1 | - | 80 | 151 | |||
| HF586902.1 | - | 80 | 151 | |||
| HF938100.1 | - | 80 | 151 | |||
| HF938101.1 | - | 80 | 151 | |||
| HF938102.1 | - | 80 | 151 | |||
| HF938103.1 | - | 80 | 151 | |||
| HF938104.1 | - | 80 | 151 | |||
| HF938105.1 | - | 80 | 151 | |||
| HG965198.1 | - | 80 | 151 | |||
| JU323311.1 | - | 154 | 151 | |||
| L27739.1 | - | 80 | 151 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

MAMU-DRA
Genomic Map