Assay Details
| Gene Symbol: GUSBA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | JU323755.1 | - | 1167 | 86 | ||
| JU334207.1 | - | 1167 | 86 | |||
| JU334208.1 | - | 1167 | 86 | |||
| JU472015.1 | - | 1178 | 86 | |||
| JU472016.1 | - | 1178 | 86 | |||
| JV649525.1 | - | 1429 | 86 | |||
| Gene Symbol: LOC677692 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015133389.2 | XP_014988875.1 | 86 | |||
| XM_028845725.1 | XP_028701558.1 | 86 | ||||
| XM_028845726.1 | XP_028701559.1 | 86 | ||||
More Information
Set Membership: |
|

GUSBA
Genomic Map