Assay Details
| Gene Symbol: MYH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851427.1 | XP_028707260.1 | 3955 | 71 | ||
| XM_028851428.1 | XP_028707261.1 | 3901 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MYH6| Gene Symbol: MYH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028851427.1 | XP_028707260.1 | 3955 | 71 | ||
| XM_028851428.1 | XP_028707261.1 | 3901 | 71 | |||
Set Membership: |
|