Assay Details
| Gene Symbol: METAP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266460.1 | NP_001253389.1 | 398 | 77 | ||
| XM_015152393.2 | XP_015007879.1 | 978 | 77 | |||
| GenBank mRNA | JU321797.1 | - | 414 | 77 | ||
| JU474104.1 | - | 388 | 77 | |||
| JV043585.1 | - | 398 | 77 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

METAP2
Genomic Map