Assay Details
| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119123.2 | XP_014974609.1 | 283 | 71 | ||
| XM_015119124.2 | XP_014974610.1 | 283 | 71 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GDPD1| Gene Symbol: GDPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015119123.2 | XP_014974609.1 | 283 | 71 | ||
| XM_015119124.2 | XP_014974610.1 | 283 | 71 | |||
Set Membership: |
|