Assay Details
| Gene Symbol: SUPT3H | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266569.1 | NP_001253498.1 | 862 | 76 | ||
| XM_015136391.2 | XP_014991877.1 | 1027 | 76 | |||
| XM_015136392.2 | XP_014991878.1 | 893 | 76 | |||
| XM_028846735.1 | XP_028702568.1 | 895 | 76 | |||
| XM_028846736.1 | XP_028702569.1 | 819 | 76 | |||
| XM_028846737.1 | XP_028702570.1 | 1022 | 76 | |||
| XM_028846738.1 | XP_028702571.1 | 1125 | 76 | |||
| XM_028846739.1 | XP_028702572.1 | 823 | 76 | |||
| XM_028846740.1 | XP_028702573.1 | 1061 | 76 | |||
| GenBank mRNA | JU336758.1 | - | 908 | 76 | ||
| JU477522.1 | - | 964 | 76 | |||
| JV048467.1 | - | 862 | 76 | |||
| JV523197.1 | - | 916 | 76 | |||
More Information
Set Membership: |
|

SUPT3H
Genomic Map