Assay Details
| Gene Symbol: RAB43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015130141.2 | XP_014985627.1 | 535 | 98 | ||
| GenBank mRNA | JU473025.1 | - | 376 | 98 | ||
| JV044166.1 | - | 376 | 98 | |||
More Information
Set Membership: |
|

RAB43
Genomic Map