Assay Details
| Gene Symbol: CNR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032825.1 | NP_001027997.1 | 1273 | 90 | ||
| GenBank mRNA | AF286025.1 | - | 1273 | 90 | ||
| JU320123.1 | - | 1556 | 90 | |||
| JV601001.1 | - | 1586 | 90 | |||
More Information
Set Membership: |
|

CNR1
Genomic Map