Assay Details
| Gene Symbol: TIMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028836870.1 | XP_028692703.1 | 767 | 85 | ||
| XM_028836871.1 | XP_028692704.1 | 453 | 85 | |||
| XM_028836872.1 | XP_028692705.1 | 423 | 85 | |||
| XM_028836873.1 | XP_028692706.1 | 359 | 85 | |||
| GenBank mRNA | AF366398.1 | - | 100 | 85 | ||
| JU328400.1 | - | 456 | 85 | |||
| JU470785.1 | - | 474 | 85 | |||
| JV043751.1 | - | 526 | 85 | |||
| JV660079.1 | - | 504 | 85 | |||
More Information
Set Membership: |
|

TIMP2
Genomic Map