Assay Details
| Gene Symbol: GREM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032848.1 | NP_001028020.1 | 250 | 128 | ||
| XM_015141773.2 | XP_014997259.2 | 433 | 128 | |||
| GenBank mRNA | AF439783.1 | - | 250 | 128 | ||
More Information
Set Membership: |
|

GREM1
Genomic Map