Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001110255.1 | NP_001103725.1 | 114 | 139 | ||
| GenBank mRNA | AF466346.1 | - | 114 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001110255.1 transcripts.
Set Membership: |
|

SPAG11B
Genomic Map