Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037282.2 | NP_001032359.1 | 580 | 103 | ||
| NM_001110255.1 | NP_001103725.1 | 182 | 103 | |||
| NM_001110259.1 | NP_001103729.1 | 394 | 103 | |||
| NM_001110260.1 | NP_001103730.1 | 502 | 103 | |||
| NM_001110261.1 | NP_001103731.1 | 453 | 103 | |||
| NM_001110262.1 | NP_001103732.1 | 433 | 103 | |||
| NM_001110263.1 | NP_001103733.1 | 374 | 103 | |||
| GenBank mRNA | AF466346.1 | - | 182 | 103 | ||
| AF466350.1 | - | 394 | 103 | |||
| AF466351.1 | - | 580 | 103 | |||
| AF466352.1 | - | 502 | 103 | |||
| AF466353.1 | - | 453 | 103 | |||
| AF466354.1 | - | 433 | 103 | |||
| AF466355.1 | - | 374 | 103 | |||
| AY528233.1 | - | 153 | 103 | |||
| AY528237.1 | - | 368 | 103 | |||
| AY528238.1 | - | 554 | 103 | |||
| AY528239.1 | - | 427 | 103 | |||
More Information
Set Membership: |
|

SPAG11B
Genomic Map