Assay Details
| Gene Symbol: ATP5F1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032901.2 | NP_001028073.1 | 119 | 72 | ||
| GenBank mRNA | AY341883.1 | - | 119 | 72 | ||
| JV045620.1 | - | 122 | 72 | |||
| JV602644.1 | - | 142 | 72 | |||
| JV635698.1 | - | 93 | 72 | |||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001032901.2 transcripts.
Set Membership: |
|

ATP5F1E
Genomic Map