Assay Details
| Gene Symbol: GRPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278448.2 | NP_001265377.1 | 733 | 57 | ||
| GenBank mRNA | AY350449.1 | - | 45 | 57 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GRPR| Gene Symbol: GRPR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001278448.2 | NP_001265377.1 | 733 | 57 | ||
| GenBank mRNA | AY350449.1 | - | 45 | 57 | ||
Set Membership: |
|