Assay Details
| Gene Symbol: KISS1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125638.2 | XP_014981124.1 | 1080 | 60 | ||
| GenBank mRNA | AY823261.1 | - | 96 | 60 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

KISS1R| Gene Symbol: KISS1R | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015125638.2 | XP_014981124.1 | 1080 | 60 | ||
| GenBank mRNA | AY823261.1 | - | 96 | 60 | ||
Set Membership: |
|