Assay Details
| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032838.1 | NP_001028010.1 | 436 | 74 | ||
| GenBank mRNA | AB035139.1 | - | 394 | 74 | ||
| AF344856.1 | - | 436 | 74 | |||
| AF530076.1 | - | 69 | 74 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

FASLG| Gene Symbol: FASLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032838.1 | NP_001028010.1 | 436 | 74 | ||
| GenBank mRNA | AB035139.1 | - | 394 | 74 | ||
| AF344856.1 | - | 436 | 74 | |||
| AF530076.1 | - | 69 | 74 | |||