Assay Details
| Gene Symbol: MICB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114956.1 | NP_001108428.1 | 335 | 120 | ||
| GenBank mRNA | AF055388.1 | - | 323 | 120 | ||
| AF322860.1 | - | 323 | 120 | |||
More Information
Set Membership: |
|

MICB
Genomic Map