Assay Details
| Gene Symbol: ABO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015116452.2 | XP_014971938.2 | 476 | 118 | ||
| XM_015116454.2 | XP_014971940.2 | 321 | 118 | |||
| GenBank mRNA | AF071830.1 | - | 81 | 118 | ||
| JV503238.1 | - | 242 | 118 | |||
| JV578799.1 | - | 116 | 118 | |||
More Information
Set Membership: |
|

ABO
Genomic Map