Assay Details
| Gene Symbol: CST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038661.1 | NP_001033750.1 | 258 | 111 | ||
| GenBank mRNA | AF218043.1 | - | 258 | 111 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CST11| Gene Symbol: CST11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001038661.1 | NP_001033750.1 | 258 | 111 | ||
| GenBank mRNA | AF218043.1 | - | 258 | 111 | ||
Set Membership: |
|