Assay Details
| Gene Symbol: IFNGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266300.2 | NP_001253229.1 | 950 | 154 | ||
| XM_015137238.2 | XP_014992724.2 | 938 | 154 | |||
| XM_028846596.1 | XP_028702429.1 | 810 | 154 | |||
| XM_028846597.1 | XP_028702430.1 | 841 | 154 | |||
| GenBank mRNA | AF227551.1 | - | 387 | 154 | ||
| JU334343.1 | - | 945 | 154 | |||
| JU475441.1 | - | 956 | 154 | |||
| JV046958.1 | - | 954 | 154 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

IFNGR1
Genomic Map