Assay Details
| Gene Symbol: STX1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032865.1 | NP_001028037.1 | 215 | 106 | ||
| GenBank mRNA | AF240771.1 | - | 215 | 106 | ||
| JV518189.1 | - | 262 | 106 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

STX1A
Genomic Map