Assay Details
| Gene Symbol: FGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028848761.1 | XP_028704594.1 | 643 | 81 | ||
| GenBank mRNA | AF251270.1 | - | 57 | 81 | ||
| JV574737.1 | - | 240 | 81 | |||
More Information
Set Membership: |
|

FGF2
Genomic Map