Assay Details
| Gene Symbol: PPIF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001266124.1 | NP_001253053.1 | 292 | 70 | ||
| GenBank mRNA | AF252388.1 | - | 89 | 70 | ||
| JU326959.1 | - | 93 | 70 | |||
| JU473867.1 | - | 269 | 70 | |||
| JV045638.1 | - | 292 | 70 | |||
| JV637471.1 | - | 285 | 70 | |||
More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
|

PPIF
Genomic Map