Assay Details
| Gene Symbol: IL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032834.2 | NP_001028006.1 | 358 | 143 | ||
| XM_015115661.2 | XP_014971147.1 | 1304 | 143 | |||
| XM_015115662.2 | XP_014971148.1 | 1393 | 143 | |||
| XM_015115663.2 | XP_014971149.1 | 1292 | 143 | |||
| GenBank mRNA | AF303732.1 | - | 358 | 143 | ||
| JV044311.1 | - | 440 | 143 | |||
| JV639328.1 | - | 461 | 143 | |||
More Information
Set Membership: |
|

IL18
Genomic Map