Assay Details
| Gene Symbol: SPINLW1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032841.1 | NP_001028013.1 | 456 | 60 | ||
| GenBank mRNA | AF346414.1 | - | 456 | 60 | ||
| JV656562.1 | - | 132 | 60 | |||
| Gene Symbol: WFDC8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_028828449.1 | XP_028684282.1 | 838 | 60 | ||
More Information
Set Membership: |
|

SPINLW1
Genomic Map