Assay Details
| Gene Symbol: CCL5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032850.1 | NP_001028022.1 | 77 | 52 | ||
| XM_015119022.2 | XP_014974508.1 | 165 | 52 | |||
| GenBank mRNA | AF449268.1 | - | 77 | 52 | ||
| DQ913691.1 | - | 165 | 52 | |||
| DQ913692.1 | - | 164 | 52 | |||
| DQ913693.1 | - | 164 | 52 | |||
| DQ913694.1 | - | 165 | 52 | |||
| DQ913695.1 | - | 164 | 52 | |||
| DQ913696.1 | - | 164 | 52 | |||
| DQ913697.1 | - | 165 | 52 | |||
| DQ913698.1 | - | 164 | 52 | |||
| DQ913699.1 | - | 165 | 52 | |||
| DQ913700.1 | - | 164 | 52 | |||
| DQ913701.1 | - | 165 | 52 | |||
| DQ913702.1 | - | 165 | 52 | |||
| DQ913703.1 | - | 165 | 52 | |||
| DQ913704.1 | - | 84 | 52 | |||
| DQ913705.1 | - | 163 | 52 | |||
| DQ913706.1 | - | 89 | 52 | |||
| DQ913707.1 | - | 90 | 52 | |||
| DQ913708.1 | - | 165 | 52 | |||
| DQ913709.1 | - | 165 | 52 | |||
| DQ913711.1 | - | 165 | 52 | |||
| DQ913712.1 | - | 83 | 52 | |||
| DQ913713.1 | - | 83 | 52 | |||
| DQ913714.1 | - | 84 | 52 | |||
| DQ913715.1 | - | 165 | 52 | |||
| DQ913716.1 | - | 164 | 52 | |||
| DQ913717.1 | - | 164 | 52 | |||
| DQ913718.1 | - | 165 | 52 | |||
| DQ913719.1 | - | 165 | 52 | |||
| DQ913721.1 | - | 164 | 52 | |||
| DQ913722.1 | - | 165 | 52 | |||
| DQ913724.1 | - | 164 | 52 | |||
| DQ913725.1 | - | 162 | 52 | |||
| DQ913726.1 | - | 164 | 52 | |||
| DQ913727.1 | - | 165 | 52 | |||
| DQ913728.1 | - | 113 | 52 | |||
| DQ913729.1 | - | 165 | 52 | |||
| DQ913730.1 | - | 164 | 52 | |||
| DQ913731.1 | - | 90 | 52 | |||
| DQ913732.1 | - | 165 | 52 | |||
| DQ913733.1 | - | 162 | 52 | |||
| JU472169.1 | - | 121 | 52 | |||
More Information
Set Membership: |
|

CCL5
Genomic Map