Assay Details
| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032877.1 | NP_001028049.1 | 67 | 89 | ||
| GenBank mRNA | AF449279.1 | - | 67 | 89 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CCL28| Gene Symbol: CCL28 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032877.1 | NP_001028049.1 | 67 | 89 | ||
| GenBank mRNA | AF449279.1 | - | 67 | 89 | ||
Set Membership: |
|