Assay Details
| Gene Symbol: CXCL13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001032880.1 | NP_001028052.1 | 271 | 75 | ||
| GenBank mRNA | AF449284.1 | - | 271 | 75 | ||
| AY288834.1 | - | 148 | 75 | |||
| JU478908.1 | - | 81 | 75 | |||
More Information
Set Membership: |
|

CXCL13
Genomic Map