Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037282.2 | NP_001032359.1 | 80 | 74 | ||
| NM_001110256.1 | NP_001103726.1 | 80 | 74 | |||
| NM_001110258.1 | NP_001103728.1 | 80 | 74 | |||
| NM_001110259.1 | NP_001103729.1 | 80 | 74 | |||
| NM_001110260.1 | NP_001103730.1 | 80 | 74 | |||
| NM_001110261.1 | NP_001103731.1 | 80 | 74 | |||
| NM_001110262.1 | NP_001103732.1 | 80 | 74 | |||
| NM_001110263.1 | NP_001103733.1 | 80 | 74 | |||
| XM_028852101.1 | XP_028707934.1 | 547 | 74 | |||
| GenBank mRNA | AF466347.1 | - | 80 | 74 | ||
| AF466349.1 | - | 80 | 74 | |||
| AF466350.1 | - | 80 | 74 | |||
| AF466351.1 | - | 80 | 74 | |||
| AF466352.1 | - | 80 | 74 | |||
| AF466353.1 | - | 80 | 74 | |||
| AF466354.1 | - | 80 | 74 | |||
| AF466355.1 | - | 80 | 74 | |||
| AY528234.1 | - | 54 | 74 | |||
| AY528236.1 | - | 54 | 74 | |||
| AY528237.1 | - | 54 | 74 | |||
| AY528238.1 | - | 54 | 74 | |||
| AY528239.1 | - | 54 | 74 | |||

SPAG11B
Genomic Map